Learn More
Invitrogen™ MFN1 Recombinant Rabbit Monoclonal Antibody (24GB4970)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557478
 View more versions of this product
Product Code. 30275289
  • £488.00 / 100µL
Description
MFN1 Recombinant Monoclonal Antibody for Western Blot, Flow
Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MFN1
Recombinant Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q811U4, Q8IWA4, Q8R4Z9
MFN1
A synthesized peptide derived from Rat Mitofusin 1 (aa 116-135).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4970
Unconjugated
MFN1
2310002F04Rik; 6330416C07Rik; D3Ertd265e; DKFZp762F247; EC 3.6.5; FLJ20693; Fzo homolog; Fzo1b; hfzo1; hfzo2; HR2; Mfn1; MFN2; MGC41806; mitochondrial mitofusin 1; mitochondrial transmembrane GTPase Fzo-1; mitochondrial transmembrane GTPase FZO1B; mitochondrial transmembrane GTPase FZO-2; mitofusin 1; mitofusin 2; mitofusin-1; mKIAA4032; putative transmembrane GTPase; transmembrane GTPase MFN1
Rabbit
Affinity chromatography
RUO
192647, 55669, 67414
-20°C