missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters

Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.

Brand:  METTLER TOLEDO™ 30207877

Code : NEW

Additional Details : Weight : 2.84000kg

 View more versions of this product

Product Code. 15375083

  • £855.00 / Each
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Instrument, LabX direct pH, USB cable, electrode clip, InLab Expert Pro-ISM-IP67, buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo™ carrying case, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.

Description

Description

  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file
  • EasyDirect software available for data transfer (model dependent)
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters

Certifications

ISO 9001, ISO 14001

Specifications

Specifications

±0.5%
±0.002
4 x AA or 1.3 V NiMH acc.
ISO 9001, ISO 14001
2000 (GLP conform)
IP 67
-2 to 20
-5°C to +130°C (ATC), -30°C to +130°C (MTC)
InLab Expert Go-ISM pH/temperature electrode; uGo carrying/protective case
Single channel; pH, Ion, ORP
0.1, 1
0.1°C
290 g
±0.1
200 to 250 hr.
5
Micro-USB (USB Device)
LCD
-2000 to 2000 mV
0.001/0.01/0.1
Wide range of samples, also suited for emulsions and suspensions; ideal for measurements on the go.
BNC (1012 Ohm)/RCA (cinch)/2mm banana
Seven2Go Pro S8 pH/mV and Ion Meter: Field Kit
±0.2°C if T: 0° to 105°C; ±0.5°C if T<0°C or T>105°C
222 x 70 x 35mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers