missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ SevenExcellence™ S475 pH/Conductivity/ISE Benchtop Meter

Product Code. 13420689 Shop All Mettler Toledo Products
Change view
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
13420689 -2 to 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13420689 Supplier METTLER TOLEDO™ Supplier No. 30046255

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The S475 meter performs pH, conductivity, resistivity, ion selective, salinity, TDS, Redox/ORP, and ISFET measurements. It can be expanded with an additional module to measure DO/BOD. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel, or LIMS system.

  • pH, conductivity, resistivity, ISE, salinity, TDS, Redox/ORP, and ISFET meter than can be expanded with an additional measurement module to measure dissolved oxygen and BOD
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Conductivity) ±0.5%
Accuracy (pH) ±0.002
Channels Multi-channel
Display Type TFT
Range (mV) -2000 to 2000 mV
Range (pH) -2 to 20
Range (Resistivity) 0.01 to 100 Mohm/cm
Range (Salinity) 0 to 80 psu, ppt
Range (TDS) 0.001 mg/L to 1000 g/L
Resolution (pH) 0.001/0.01/0.1
Temperature Range (Metric) -30°C to +130°C
Includes S475 pH/conductivity/ISE meter, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, 2 buffer sachets each for pH 1413 mS/cm and 12.88 mS/cm, test certificate
Languages Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 250 analyses (20.000 data points)
Model S475 pH/Conductivity/ISE Meter
Range (ISE) 0.001 to 999999mg/L ppm, 0.0001 to 100mol/L %, 0.001 to 100000mmol/L
Resolution (Conductivity) 0.001μS/cm to 2000mS/cm
Resolution (mV) 0.1/1mV
Temperature Accuracy ±0.1°C
Temperature Resolution 0.1°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.