missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ SevenCompact™ S220 pH Benchtop Meter

Product Code. 15937765 Shop All Mettler Toledo Products
Change view
Click to view available options
Model:
Automatic Stirring Option
Bio/Life Science Option
General Applications Option
Micro Option
S220 Basic pH Meter
Water/Beverage Option
Unit Size:
1 set
1 unit
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15937765 Automatic Stirring Option Each
11374233 S220 Basic pH Meter Each
11304233 Bio/Life Science Option Each
11384233 General Applications Option 1 set
11344233 Water/Beverage Option Each
15573423 Micro Option 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15937765 Supplier METTLER TOLEDO™ Supplier No. 30096986

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The S220 meter measures pH, ion concentration, and Redox/ORP. A simple and durable interface with only 2 buttons makes this meter very easy to operate, and results can be captured by a printer with a time and date stamp. Available as meter only or ready-to-use kits for specific applications.

  • pH, Ion Concentration, and Redox/ORP benchtop meter that can display results in pH, mV, and Rel mV
  • Durable and simple to operate with two buttons: 'Read' and 'Cal'
  • IP54 Protection
  • Comes with electrode arm that moves straight up and down, bringing the electrode into position where it works best for your sample
  • Compatible with a wide range of electrodes for any application
  • Compatible with ISM electrodes that capture and save calibration data, which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Results can be captured by a printer with a time and date stamp
  • Optional installation and qualification (IQ/OQ) package available through METTLER TOLEDO Service
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ±0.1
Accuracy (pH) ±0.002
Description S220 Kit - General Applications + Automatic Stirring
Channels 1
Display Type High-resolution Color
Range (mV) -2000.0 to 2000.0
Range (pH) -2 to 20
Resolution (pH) 0.001/0.01/0.1
Temperature Range (Metric) -30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC)
For Use With (Application) General Applications, Automatic Stirring
Includes S220 pH meter, uMix magnetic stirrer, InLab Expert Pro-ISM electrode, electrode holder, 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01, pH Measurement Guide, protective cover, test certificate
Memory 1.000 measurements
Model Automatic Stirring Option
Product Type pH/Ion Benchtop Meter
Resolution (mV) 0.1/1mV
Temperature Accuracy ±0.1°C
Temperature Range (English) MTC: -86°F to +266°F, ATC: -41°F to +266°F
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.