missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ SevenCompact™ S220 pH Benchtop Meter

Product Code. 11344233 Shop All Mettler Toledo Products
Change view
Click to view available options
Model:
Automatic Stirring Option
Bio/Life Science Option
General Applications Option
Micro Option
S220 Basic pH Meter
Water/Beverage Option
Unit Size:
1 set
1 unit
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
11344233 Water/Beverage Option Each
11374233 S220 Basic pH Meter Each
11304233 Bio/Life Science Option Each
11384233 General Applications Option 1 set
15937765 Automatic Stirring Option Each
15573423 Micro Option 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11344233 Supplier METTLER TOLEDO™ Supplier No. 30019032

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The S220 meter measures pH, ion concentration, and Redox/ORP. A simple and durable interface with only 2 buttons makes this meter very easy to operate, and results can be captured by a printer with a time and date stamp. Available as meter only or ready-to-use kits for specific applications.

  • pH, Ion Concentration, and Redox/ORP benchtop meter that can display results in pH, mV, and Rel mV
  • Durable and simple to operate with two buttons: 'Read' and 'Cal'
  • IP54 Protection
  • Comes with electrode arm that moves straight up and down, bringing the electrode into position where it works best for your sample
  • Compatible with a wide range of electrodes for any application
  • Compatible with ISM electrodes that capture and save calibration data, which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Results can be captured by a printer with a time and date stamp
  • Optional installation and qualification (IQ/OQ) package available through METTLER TOLEDO Service
TRUSTED_SUSTAINABILITY

Specifications

Outputs USB, RS-232
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Calibration Points 5
Data Management Details 1000 Measurements
Description S220 Kit - Water & Beverage Measurements
Display Type TFT
Range (mV) -2000 to 2000 mV
Range (pH) -2 to 20
Resolution (pH) 0.001/0.01/0.1
Temperature Range (Metric) -30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC)
For Use With (Application) Water & Beverage
Includes S220 pH meter, InLab Versatile Pro electrode, electrode holder, 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01, pH Measurement Guide, protective cover, test certificate
Model Water/Beverage Option
Probe Type pH/Ion
Product Type Meter Kit: S220-U
Resolution (mV) 0.1/1mV
Temperature Accuracy ±0.1°C
Temperature Range (English) MTC: -86°F to +266°F, ATC: -41°F to +266°F
Voltage 9/12 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.