Learn More
METTL7A Mouse anti-Human, Unconjugated, Clone: 1A9B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67905-1-IG-150UL
Description
The METTL7A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human METTL7A. This antibody recognizes human, mouse, rat, rabbit antigen. The METTL7A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| METTL7A | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 25840 | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 1A9B4 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit |