missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL7A Mouse anti-Human, Unconjugated, Clone: 1A9B4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | METTL7A |
|---|---|
| Clone | 1A9B4 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17442403
|
Proteintech
67905-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17432403
|
Proteintech
67905-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The METTL7A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human METTL7A. This antibody recognizes human, mouse, rat, rabbit antigen. The METTL7A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| METTL7A | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Mouse, Rat, Rabbit | |
| IgG1 |
| 1A9B4 | |
| Monoclonal | |
| Mouse | |
| 25840 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title