missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL3 Rabbit anti-Human, Unconjugated, Clone: 2I9, Proteintech
Rabbit Recombinant Antibody
£135.00 - £360.00
Specifications
| Antigen | METTL3 |
|---|---|
| Clone | 2I9 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17482453
|
Proteintech
80323-1-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17472453
|
Proteintech
80323-1-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The METTL3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human METTL3. This antibody recognizes human, mouse, rat antigen. The METTL3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| METTL3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 2I9 | |
| Monoclonal | |
| Rabbit | |
| 56339 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title