Learn More
METTL3 Rabbit anti-Human, Unconjugated, Clone: 2I9, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80323-1-RR-20UL
Description
The METTL3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human METTL3. This antibody recognizes human, mouse, rat antigen. The METTL3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| METTL3 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 56339 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2I9 | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |