Learn More
Metoprolol tartrate, 98+%, Thermo Scientific™
A beta-adrenergic blocker
£62.65 - £269.00
Chemical Identifiers
| CAS | 56392-17-7 |
|---|---|
| Molecular Formula | C34H56N2O12 |
| Molecular Weight (g/mol) | 684.824 |
| MDL Number | MFCD00056257 |
| InChI Key | YGULWPYYGQCFMP-NPHUUBOGSA-N |
| Synonym | y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2 |
| PubChem CID | 11957594 |
| IUPAC Name | (2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
| SMILES | CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15444409
|
Thermo Scientific Alfa Aesar
J61920.06 |
5g |
£69.20 £62.65 / 5g Save £6.55 9% Off |
Estimated Shipment: 22-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15454409
|
Thermo Scientific Alfa Aesar
J61920.14 |
25g |
£269.00
25g |
Estimated Shipment: 06-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMetoprolol tartrate is an antagonist of the β1-AR (β1-adrenoceptor). It inhibits spontaneous endothelin-1 production in vitro and displays antihypertensive and antianginal activity in vivo.
Solubility
Soluble in water (>1000 mg/ml), methanol (>500 mg/ml), chloroform (496 mg/ml), ethanol (31 mg/ml at 25°C), and DMSO (100 mg/ml at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Chemical Identifiers
| 56392-17-7 | |
| 684.824 | |
| YGULWPYYGQCFMP-NPHUUBOGSA-N | |
| 11957594 | |
| CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O |
| C34H56N2O12 | |
| MFCD00056257 | |
| y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2 | |
| (2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
Specifications
| 56392-17-7 | |
| MFCD00056257 | |
| y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2 | |
| YGULWPYYGQCFMP-NPHUUBOGSA-N | |
| (2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol | |
| 11957594 | |
| ≥98% |
| C34H56N2O12 | |
| 14,6151 | |
| Soluble in water (>1000mg/ml),methanol (>500mg/ml),chloroform (496mg/ml),ethanol (31mg/ml at 25°C),and DMSO (100mg/ml at 25°C). | |
| CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O | |
| 684.824 | |
| 684.82 | |
| Metoprolol tartrate |
Safety and Handling
missing translation for 'einecsNumber' : 260-148-9
missing translation for 'rtecsNumber' : UB7450100
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.