missing translation for 'onlineSavingsMsg'
Learn More

Metoprolol tartrate, 98+%

Product Code. 15454409
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454409 25 g 25g
15444409 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454409 Supplier Thermo Scientific Alfa Aesar Supplier No. J61920.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A beta-adrenergic blocker | CAS: 56392-17-7 | C34H56N2O12 | 684.824 g/mol

Metoprolol tartrate is an antagonist of the β1-AR (β1-adrenoceptor). It inhibits spontaneous endothelin-1 production in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metoprolol tartrate is an antagonist of the β1-AR (β1-adrenoceptor). It inhibits spontaneous endothelin-1 production in vitro and displays antihypertensive.

Solubility
Soluble in water (>1000 mg/ml), methanol (>500 mg/ml), chloroform (496 mg/ml), ethanol (31 mg/ml at 25°C), and DMSO (100 mg/ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56392-17-7
Molecular Formula C34H56N2O12
Molecular Weight (g/mol) 684.824
MDL Number MFCD00056257
InChI Key YGULWPYYGQCFMP-NPHUUBOGSA-N
Synonym y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2
PubChem CID 11957594
IUPAC Name (2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Merck Index 14,6151
Solubility Information Soluble in water (>1000mg/ml),methanol (>500mg/ml),chloroform (496mg/ml),ethanol (31mg/ml at 25°C),and DMSO (100mg/ml at 25°C).
Formula Weight 684.82
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Metoprolol tartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.