Learn More
MedChemExpress Methylene Blue
Description
Methylene blue is a vasopressor and is often used as a dye in several medical procedures. Methylene blue through the nitric oxide syntase/guanylate cyclase signalling pathway to reduce prepulse inhibition. Methylene blue is a REDOX cycling compound and able to cross the blood-brain barrier. Methylene blue is a Tau aggregation inhibitor. Methylene blue reduces cerebral edema, attenuated microglial activation and reduced neuroinflammation.
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Quantity | 500 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 100 mg/mL (312.65 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 319.85 g/mol |
| CAS | 61-73-4 |
| Synonym | Basic Blue 9 ∣ CI-52015 ∣ Methylthioninium chloride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.