missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Methylene Blue

Product Code. 16471726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471726 500 mg 500mg
16416486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471726 Supplier MedChemExpress Supplier No. HY14536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methylene blue (Basic Blue 9) is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A) and NO synthase (NOS) inhibitor.

Methylene blue is a vasopressor and is often used as a dye in several medical procedures. Methylene blue through the nitric oxide syntase/guanylate cyclase signalling pathway to reduce prepulse inhibition. Methylene blue is a REDOX cycling compound and able to cross the blood-brain barrier. Methylene blue is a Tau aggregation inhibitor. Methylene blue reduces cerebral edema, attenuated microglial activation and reduced neuroinflammation.

Specifications

Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Quantity 500 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (312.65 mM; Need ultrasonic)
Molecular Weight (g/mol) 319.85 g/mol
CAS 61-73-4
Synonym Basic Blue 9 ∣ CI-52015 ∣ Methylthioninium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.