Learn More
MedChemExpress Methylene Blue
Description
Methylene blue is a vasopressor and is often used as a dye in several medical procedures. Methylene blue through the nitric oxide syntase/guanylate cyclase signalling pathway to reduce prepulse inhibition. Methylene blue is a REDOX cycling compound and able to cross the blood-brain barrier. Methylene blue is a Tau aggregation inhibitor. Methylene blue reduces cerebral edema, attenuated microglial activation and reduced neuroinflammation.
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 100 mg/mL (312.65 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 319.85 g/mol |
| CAS | 61-73-4 |
| Synonym | Basic Blue 9 ∣ CI-52015 ∣ Methylthioninium chloride |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.