missing translation for 'onlineSavingsMsg'
Learn More

Methyl malonamate, 98%

Product Code. 11427006
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427006 100 g 100g
11417006 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427006 Supplier Thermo Scientific Alfa Aesar Supplier No. B25326.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51513-29-2 | C4H7NO3 | 117.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51513-29-2
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.10
MDL Number MFCD00674533
InChI Key LSNSJCKGQREPDW-UHFFFAOYSA-N
Synonym methyl malonamate, methyl malonate monoamide, methyl 2-carbamoylacetate, propanoic acid, 3-amino-3-oxo-, methyl ester, acmc-20akfl, methyl carbamoylacetate, methyl-malonate monoamide, malonamic acid methyl ester, malonamidic acid methyl ester, methyl 3-amino-3-oxo-propanoate
PubChem CID 3595299
IUPAC Name methyl 3-amino-3-oxopropanoate
SMILES COC(=O)CC(N)=O

Specifications

Melting Point 38°C to 40°C
Quantity 100 g
Formula Weight 117.11
Percent Purity 98%
Chemical Name or Material Methyl malonamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.