CAS RN 51513-29-2
CAS RN 51513-29-2
IUPAC Name:
methyl 2-carbamoylacetate
Synonyms:
methyl 2-carbamoylacetate
Molecular Weight (g/mol):
117.1
Molecular Formula:
C4H7NO3
InChi Key:
LSNSJCKGQREPDW-UHFFFAOYSA-N
SMILES:
COC(=O)CC(N)=O
1
–
1
of
1
results
1
Methyl malonamate, 98%
CAS: 51513-29-2 Molecular Formula: C4H7NO3 Molecular Weight (g/mol): 117.10 MDL Number: MFCD00674533 InChI Key: LSNSJCKGQREPDW-UHFFFAOYSA-N Synonym: methyl malonamate,methyl malonate monoamide,methyl 2-carbamoylacetate,propanoic acid, 3-amino-3-oxo-, methyl ester,acmc-20akfl,methyl carbamoylacetate,methyl-malonate monoamide,malonamic acid methyl ester,malonamidic acid methyl ester,methyl 3-amino-3-oxo-propanoate PubChem CID: 3595299 IUPAC Name: methyl 3-amino-3-oxopropanoate SMILES: COC(=O)CC(N)=O