missing translation for 'onlineSavingsMsg'
Learn More

Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™

Product Code. 10103992
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103992 10 g Amber Glass Bottle 10g
10095472 1 g Amber Glass Bottle 1g
10156932 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103992 Supplier Thermo Scientific Maybridge Supplier No. KM00619EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4338-95-8
Molecular Formula C2H7IN2S
Molecular Weight (g/mol) 218.06
MDL Number MFCD00035598
InChI Key LFXAECSQJSRSTP-UHFFFAOYSA-N
Synonym methyl aminomethanimidothioate hydroiodide, sulfiode, methylthiouronium iodide, methyl carbamimidothioate hydroiodide, s-methylthiuronium iodide, s-methylthiouronium iodide, s-methylisothiouronium iodide, 2-methyl-2-thiopseudourea hydroiodide, 2-methylisothiouronium iodide, s-methylisothiourea hydriodide
PubChem CID 197812
SMILES [H+].[I-].CSC(N)=N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name methyl carbamimidothioate;hydroiodide
Formula Weight 218.06
Percent Purity 95%
Chemical Name or Material Methyl aminomethanimidothioate hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.