missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-hydroxynicotinate, 97+%, Thermo Scientific Chemicals

Product Code. 10463925 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10463925 5 g Glass bottle 5g
10608414 250 mg Glass bottle 250mg
10055193 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10463925 Supplier Thermo Scientific Acros Supplier No. 393350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30766-22-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00191521
InChI Key KJJSHOHQQHACLE-UHFFFAOYSA-N
PubChem CID 354317
IUPAC Name methyl 5-hydroxypyridine-3-carboxylate
SMILES COC(=O)C1=CC(O)=CN=C1

Specifications

Melting Point 195°C to 197°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Synonym methyl 5-hydroxynicotinate, 5-hydroxynicotinic acid methyl ester, methyl5-hydroxynicotinate, methyl 5-hydroxy-3-pyridinecarboxylate, 5-hydroxy-nicotinic acid methyl ester, methyl-5-hydroxynicotinate, 5-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 5-hydroxy-, methyl ester, 5-hydroxy nicotinic acid methyl ester, acmc-1crme
Formula Weight 153.14
Percent Purity 97+%
Physical Form Powder or Lumps
Chemical Name or Material Methyl 5-hydroxynicotinate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 5-hydroxynicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.