accessibility menu, dialog, popup

UserName

CH3OONOH

CAS RN 30766-22-4

IUPAC Name: methyl 5-hydroxypyridine-3-carboxylate
Synonyms: methyl 5-hydroxypyridine-3-carboxylate
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: KJJSHOHQQHACLE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=CN=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)