Learn More
Methyl 4-fluorobenzoate, 98%, Thermo Scientific Chemicals
£45.30 - £112.00
Chemical Identifiers
CAS | 403-33-8 |
---|---|
Molecular Formula | C8H7FO2 |
Molecular Weight (g/mol) | 154.14 |
InChI Key | MSEBQGULDWDIRW-UHFFFAOYSA-N |
Synonym | methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019 |
PubChem CID | 67878 |
IUPAC Name | methyl 4-fluorobenzoate |
SMILES | COC(=O)C1=CC=C(C=C1)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15580604
|
Thermo Scientific Acros
459240250 |
25 g |
£45.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15590604
|
Thermo Scientific Acros
459241000 |
100 g |
£112.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
403-33-8 | |
154.14 | |
methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019 | |
methyl 4-fluorobenzoate |
C8H7FO2 | |
MSEBQGULDWDIRW-UHFFFAOYSA-N | |
67878 | |
COC(=O)C1=CC=C(C=C1)F |
Specifications
403-33-8 | |
90°C to 92°C (27.0 hPa) | |
Authentic | |
C8H7FO2 | |
methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019 | |
COC(=O)C1=CC=C(C=C1)F | |
154.14 | |
154.14 | |
Methyl 4-fluorobenzoate |
1.1920g/mL | |
78°C | |
97.5% min. (GC) | |
1.192 | |
MSEBQGULDWDIRW-UHFFFAOYSA-N | |
methyl 4-fluorobenzoate | |
67878 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-956-7
- Methyl 4-fluorobenzoate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only