missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorobenzoate, 98%

Product Code. 15590604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15590604 100 g 100g
15580604 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15590604 Supplier Thermo Scientific Acros Supplier No. 459241000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 403-33-8 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-33-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
InChI Key MSEBQGULDWDIRW-UHFFFAOYSA-N
Synonym methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019
PubChem CID 67878
IUPAC Name methyl 4-fluorobenzoate
SMILES COC(=O)C1=CC=C(C=C1)F

Specifications

Density 1.1920g/mL
Boiling Point 90°C to 92°C (27.0 hPa)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 100 g
Specific Gravity 1.192
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material Methyl 4-fluorobenzoate
compliance-icons compliance-icons
Product Identifier
  • Methyl 4-fluorobenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.