missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-cyanocinnamate, 97%, Thermo Scientific™

Product Code. 11474670
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474670 250 mg 250mg
11484670 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474670 Supplier Thermo Scientific Alfa Aesar Supplier No. H50742.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52116-83-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00157182,MFCD00157182
InChI Key CDZMIJFSAZPYEZ-UHFFFAOYSA-N
Synonym methyl 4-cyanocinnamate, 2-propenoic acid, 3-4-cyanophenyl-, methyl ester, methyl 3-4-cyanophenyl prop-2-enoate, 3-4-cyano-phenyl-acrylic acid methyl ester, acmc-20ank5
PubChem CID 53412322
IUPAC Name methyl 3-(4-cyanophenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(C=C1)C#N

Specifications

Melting Point 112°C to 125°C
Quantity 250 mg
UN Number UN3439
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material Methyl 4-cyanocinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.