CAS RN 52116-83-3
IUPAC Name:
methyl 3-(4-cyanophenyl)prop-2-enoate
Synonyms:
methyl 3-(4-cyanophenyl)prop-2-enoate
Molecular Weight (g/mol):
187.2
Molecular Formula:
C11H9NO2
InChi Key:
CDZMIJFSAZPYEZ-UHFFFAOYSA-N
SMILES:
COC(=O)C=CC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
Methyl 4-cyanocinnamate, 97%, Thermo Scientific™
CAS: 52116-83-3 Molecular Formula: C11H9NO2 Molecular Weight (g/mol): 187.20 MDL Number: MFCD00157182,MFCD00157182 InChI Key: CDZMIJFSAZPYEZ-UHFFFAOYSA-N Synonym: methyl 4-cyanocinnamate,2-propenoic acid, 3-4-cyanophenyl-, methyl ester,methyl 3-4-cyanophenyl prop-2-enoate,3-4-cyano-phenyl-acrylic acid methyl ester,acmc-20ank5 PubChem CID: 53412322 IUPAC Name: methyl 3-(4-cyanophenyl)prop-2-enoate SMILES: COC(=O)C=CC1=CC=C(C=C1)C#N