missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorobenzoate, 99%

Product Code. 11419352
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419352 100 g 100g
11409352 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419352 Supplier Thermo Scientific Alfa Aesar Supplier No. A10109.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1126-46-1 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126-46-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000621
InChI Key LXNFVVDCCWUUKC-UHFFFAOYSA-N
Synonym methyl p-chlorobenzoate, 4-chlorobenzoic acid methyl ester, benzoic acid, 4-chloro-, methyl ester, unii-m0u80j80va, benzoic acid, p-chloro-, methyl ester, methyl-p-chlorobenzoate, p-chlorobenzoic acid methyl ester, methyl-4-chlorobenzoate, 4-chloro-benzoic acid methyl ester, decitabine related compound d
PubChem CID 14307
IUPAC Name methyl 4-chlorobenzoate
SMILES COC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 41°C to 45°C
Density 1.382
Boiling Point 136°C to 137°C (10 mmHg)
Flash Point 106°C (222°F)
Quantity 100 g
Beilstein 1564310
Merck Index 14,2125
Formula Weight 170.6
Percent Purity 99%
Chemical Name or Material Methyl 4-chlorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.