missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloro-3-methylbenzoate, 97%, Thermo Scientific™

Product Code. 11377896
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377896 2 g 2g
11387896 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377896 Supplier Thermo Scientific Alfa Aesar Supplier No. L13110.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91367-05-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00151838
InChI Key QOTGNXMPQNGYDM-UHFFFAOYSA-N
Synonym 4-chloro-3-methylbenzoic acid methyl ester, benzoic acid, 4-chloro-3-methyl-, methyl ester, pubchem3710, acmc-209r8w, 2-chloro-5-methoxycarbonyl toluene, methyl 4-chloranyl-3-methyl-benzoate, 4-chloro-m-toluic acid methyl ester, 4-chloro-3-methylbenzoic acid, methyl ester
PubChem CID 7015786
IUPAC Name methyl 4-chloro-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)Cl

Specifications

Refractive Index 1.537
Quantity 2 g
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material Methyl 4-chloro-3-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.