CAS RN 91367-05-4
IUPAC Name:
methyl 4-chloro-3-methylbenzoate
Synonyms:
methyl 4-chloro-3-methylbenzoate
Molecular Weight (g/mol):
184.62
Molecular Formula:
C9H9ClO2
InChi Key:
QOTGNXMPQNGYDM-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(C)=C(Cl)C=C1
1
–
1
of
1
results
1
Methyl 4-chloro-3-methylbenzoate, 97%, Thermo Scientific™
CAS: 91367-05-4 Molecular Formula: C9H9ClO2 Molecular Weight (g/mol): 184.619 MDL Number: MFCD00151838 InChI Key: QOTGNXMPQNGYDM-UHFFFAOYSA-N Synonym: 4-chloro-3-methylbenzoic acid methyl ester,benzoic acid, 4-chloro-3-methyl-, methyl ester,pubchem3710,acmc-209r8w,2-chloro-5-methoxycarbonyl toluene,methyl 4-chloranyl-3-methyl-benzoate,4-chloro-m-toluic acid methyl ester,4-chloro-3-methylbenzoic acid, methyl ester PubChem CID: 7015786 IUPAC Name: methyl 4-chloro-3-methylbenzoate SMILES: CC1=C(C=CC(=C1)C(=O)OC)Cl