missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminobutyrate hydrochloride, 99%

Product Code. 11975391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975391 25 g Glass Bottle 25g
11995391 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975391 Supplier Thermo Scientific Acros Supplier No. 442910250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13031-60-2 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-60-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00043270
InChI Key WPGPRLVPWACBHW-UHFFFAOYSA-N
Synonym methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
PubChem CID 2756383
IUPAC Name methyl 4-aminobutanoate;hydrochloride
SMILES [Cl-].COC(=O)CCC[NH3+]

Specifications

Melting Point 118°C to 119°C
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Glass Bottle
Quantity 25 g
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material Methyl 4-aminobutyrate hydrochloride
compliance-icons
Product Identifier
  • Methyl 4-aminobutyrate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.