CAS RN 13031-60-2
IUPAC Name:
4-methoxy-4-oxobutan-1-aminium chloride
Synonyms:
4-methoxy-4-oxobutan-1-aminium chloride
Molecular Weight (g/mol):
153.61
Molecular Formula:
C5H12ClNO2
InChi Key:
WPGPRLVPWACBHW-UHFFFAOYSA-N
SMILES:
[Cl-].COC(=O)CCC[NH3+]
1
–
1
of
1
results
1
Methyl 4-aminobutyrate hydrochloride, 99%
CAS: 13031-60-2 Molecular Formula: C5H12ClNO2 Molecular Weight (g/mol): 153.61 MDL Number: MFCD00043270 InChI Key: WPGPRLVPWACBHW-UHFFFAOYSA-N Synonym: methyl 4-aminobutanoate hydrochloride,methyl 4-aminobutyrate hydrochloride,methyl 4-aminobutanoate hcl,methyl 4-aminobutyrate hcl,h-gamma-abu-ome.hcl,4-aminobutyric acid methyl ester hydrochloride,4-aminobutyric acid methyl ester, hydrochloride,4-amino-n-butyric acid methyl ester hydrochloride,butanoic acid, 4-amino-, methyl ester, hydrochloride,4-amino-butyric acid methyl ester hydrochloride PubChem CID: 2756383 IUPAC Name: methyl 4-aminobutanoate;hydrochloride SMILES: [Cl-].COC(=O)CCC[NH3+]