missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-iodobenzoate, 95%, Thermo Scientific Chemicals

Product Code. 10081274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081274 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10081274 Supplier Thermo Scientific Acros Supplier No. 397070050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19718-49-1
Molecular Formula C8H8INO2
Molecular Weight (g/mol) 277.061
MDL Number MFCD00090423
InChI Key MRLVFVTVXSKAMX-UHFFFAOYSA-N
Synonym 4-amino-3-iodo-benzoic acid methyl ester, methyl 3-iodo-4-aminobenzoate, methyl 4-amino-3-iodobenzozte, 4-amino-3-iodobenzoic acid methyl ester, benzoic acid, 4-amino-3-iodo-, methyl ester, methyl 4-amino-3-iodo-benzoate, pubchem10432, acmc-1c8qj, methyl4-amino-3-iodobenzoate, methyl 4-amino-3-iodobenzoat
PubChem CID 4104649
IUPAC Name methyl 4-amino-3-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)I

Specifications

Melting Point 86.0°C to 91.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula NH2(I)C6H3COOCH3
Quantity 5 g
Formula Weight 277.06
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-amino-3-iodobenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.