accessibility menu, dialog, popup

UserName

CH3OONH2I

CAS RN 19718-49-1

IUPAC Name: methyl 4-amino-3-iodobenzoate
Synonyms: methyl 4-amino-3-iodobenzoate
Molecular Weight (g/mol): 277.06
Molecular Formula: C8H8INO2
InChi Key: MRLVFVTVXSKAMX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(I)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results