missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-hydroxybenzoate, 98%

Product Code. 11377545
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377545 10 g 10g
11367545 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377545 Supplier Thermo Scientific Alfa Aesar Supplier No. L05687.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63435-16-5 | C8H9NO3 | 167.164 g/mol

Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63435-16-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00017093
InChI Key OCZXDVNSNDITBS-UHFFFAOYSA-N
Synonym methyl-4-amino-3-hydroxybenzoate, benzoic acid, 4-amino-3-hydroxy-, methyl ester, methyl 4-amino-3-hydroxy-benzoate, methyl 4-amino-3-hydroxybenzenecarboxylate, methyl4-amino-3-hydroxybenzoate, pubchem3149, acmc-1b7ml, 2-amino-5-methoxycarbonylphenol, methyl3-hydroxy-4-aminobenzoate, 2-hydroxy 4-carbomethoxy aniline
PubChem CID 7020619
IUPAC Name methyl 4-amino-3-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)O

Specifications

Melting Point 118°C to 122°C
Quantity 10 g
Beilstein 608716
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material Methyl 4-amino-3-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.