accessibility menu, dialog, popup

UserName

CAS RN 63435-16-5

IUPAC Name: methyl 3-amino-4-hydroxybenzoate
Synonyms: orthoform orthoderm orthocaine 奥索卡因
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: VNQABZCSYCTZMS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results