missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-hydroxybenzoate, 98%

Product Code. p-7058305
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11367545

Brand: Thermo Scientific Alfa Aesar L05687.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63435-16-5 | C8H9NO3 | 167.164 g/mol

Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63435-16-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00017093
InChI Key OCZXDVNSNDITBS-UHFFFAOYSA-N
Synonym methyl-4-amino-3-hydroxybenzoate, benzoic acid, 4-amino-3-hydroxy-, methyl ester, methyl 4-amino-3-hydroxy-benzoate, methyl 4-amino-3-hydroxybenzenecarboxylate, methyl4-amino-3-hydroxybenzoate, pubchem3149, acmc-1b7ml, 2-amino-5-methoxycarbonylphenol, methyl3-hydroxy-4-aminobenzoate, 2-hydroxy 4-carbomethoxy aniline
PubChem CID 7020619
IUPAC Name methyl 4-amino-3-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)O

Specifications

Melting Point 118°C to 122°C
Quantity 2 g
Beilstein 608716
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material Methyl 4-amino-3-hydroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.