missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-phenylpropionate, 98%

Product Code. 11367385
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367385 50 g 50g
11377385 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367385 Supplier Thermo Scientific Alfa Aesar Supplier No. L05310.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-25-3 | C10H12O2 | 164.204 g/mol

Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-25-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00017209
InChI Key RPUSRLKKXPQSGP-UHFFFAOYSA-N
Synonym methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
PubChem CID 7643
IUPAC Name methyl 3-phenylpropanoate
SMILES COC(=O)CCC1=CC=CC=C1

Specifications

Density 1.045
Boiling Point 236°C to 239°C
Flash Point 100°C (212°F)
Refractive Index 1.502
Quantity 50 g
Beilstein 1909591
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Methyl 3-phenylpropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.