missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxy-4-nitrobenzoate, 98%

Product Code. 11430797
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11430797

Brand: Thermo Scientific Alfa Aesar B23196.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5081-37-8 | C9H9NO5 | 211.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5081-37-8
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00051967
InChI Key PVVFEPFLFHDHHK-UHFFFAOYSA-N
Synonym 3-methoxy-4-nitrobenzoic acid methyl ester, benzoic acid, 3-methoxy-4-nitro-, methyl ester, benzoic acid, 5-methoxy-4-nitro-, methyl ester, methyl3-methoxy-4-nitrobenzoate, pubchem4216, acmc-209kq5, ksc275c3p, methylmethoxynitrobenzenecarboxylate, 4-nitro-3-methoxy benzoic acid, 3-methoxy-4-nitro-benzoic acid methyl ester
PubChem CID 287308
IUPAC Name methyl 3-methoxy-4-nitrobenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]

Specifications

Melting Point 83°C to 87°C
Quantity 5 g
Beilstein 2856207
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxy-4-nitrobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.