accessibility menu, dialog, popup

UserName

CAS RN 5081-37-8

CH3OOOH3CNOO

CAS RN 5081-37-8

IUPAC Name: methyl 3-methoxy-4-nitrobenzoate
Synonyms: methyl 3-methoxy-4-nitrobenzoate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: PVVFEPFLFHDHHK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C(OC)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results