missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chlorobenzo[b]thiophene-2-carboxylate, 95%, Thermo Scientific™

Product Code. p-7045430
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382936 1 g 1g
11392936 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382936 Supplier Thermo Scientific Alfa Aesar Supplier No. L10696.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21211-07-4
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.674
MDL Number MFCD00052503
InChI Key VAPMGETZRJYSLW-UHFFFAOYSA-N
Synonym methyl 3-chlorobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-chloro-, methyl ester, maybridge1_000462, acmc-1co9m, methyl 3-chlorobenzothiophene-2-carboxylate, methyl 3-chloranyl-1-benzothiophene-2-carboxylate, 3-chloro-1-benzothiophene-2-carboxylic acid methyl ester
PubChem CID 607073
IUPAC Name methyl 3-chloro-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=C(C2=CC=CC=C2S1)Cl

Specifications

Melting Point 80°C to 82°C
Odor Odorless
Quantity 1 g
Beilstein 1312686
Formula Weight 226.68
Percent Purity 95%
Chemical Name or Material Methyl 3-chlorobenzo[b]thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.