accessibility menu, dialog, popup

UserName

CAS RN 21211-07-4

H3COOSCl

CAS RN 21211-07-4

IUPAC Name: methyl 3-chloro-1-benzothiophene-2-carboxylate
Synonyms: methyl 3-chloro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 226.67
Molecular Formula: C10H7ClO2S
InChi Key: VAPMGETZRJYSLW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results