missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-6-iodopyrazine-2-carboxylate, 96%, Thermo Scientific™

Product Code. 15408106
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408106 250 mg 250mg
15489458 1 g 1g
15499458 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408106 Supplier Thermo Scientific Alfa Aesar Supplier No. H63026.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1458-16-8
Molecular Formula C6H6IN3O2
Molecular Weight (g/mol) 279.037
MDL Number MFCD09909640
InChI Key FDLARAKNPMCCKJ-UHFFFAOYSA-N
Synonym 3-amino-6-iodopyrazine-2-carboxylic acid methyl ester, 3-amino-6-iodo-pyrazine-2-carboxylic acid methyl ester, methyl 3-amino-6-iodo-2-pyrazinecarboxylate, methyl 3-azanyl-6-iodanyl-pyrazine-2-carboxylate, 3-amino-6-iodo-2-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid,3-amino-6-iodo-,methyl ester, 2-pyrazinecarboxylicacid, 3-amino-6-iodo-, methyl ester
PubChem CID 11087124
IUPAC Name methyl 3-amino-6-iodopyrazine-2-carboxylate
SMILES COC(=O)C1=NC(=CN=C1N)I

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 279.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 3-amino-6-iodopyrazine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.