accessibility menu, dialog, popup

UserName

CH3OONNH2NI

CAS RN 1458-16-8

IUPAC Name: methyl 3-amino-6-iodopyrazine-2-carboxylate
Synonyms: methyl 3-amino-6-iodopyrazine-2-carboxylate
Molecular Weight (g/mol): 279.04
Molecular Formula: C6H6IN3O2
InChi Key: FDLARAKNPMCCKJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(I)=CN=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results