missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-dimethylacrylate, 98%

Product Code. 11305585
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11305585

Brand: Thermo Scientific Alfa Aesar L19731.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 924-50-5 | C6H10O2 | 114.144 g/mol

Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in chloroform. Insoluble in water,

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-50-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00043940
InChI Key FZIBCCGGICGWBP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate
PubChem CID 13546
IUPAC Name methyl 3-methylbut-2-enoate
SMILES CC(=CC(=O)OC)C

Specifications

Melting Point -41°C
Density 0.936
Boiling Point 134°C to 139°C
Flash Point 33°C (91°F)
Linear Formula (CH3)2C=CHCO2CH3
Refractive Index 1.4375
Quantity 10 g
UN Number UN3272
Beilstein 1741592
Sensitivity Light sensitive
Solubility Information Soluble in chloroform. Insoluble in water,
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material Methyl 3,3-dimethylacrylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.