CAS RN 924-50-5
CAS RN 924-50-5
IUPAC Name:
methyl 3-methylbut-2-enoate
Synonyms:
methyl 3-methylbut-2-enoate
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
FZIBCCGGICGWBP-UHFFFAOYSA-N
SMILES:
COC(=O)C=C(C)C
1
–
1
of
1
results
1
Methyl 3,3-dimethylacrylate, 98%
CAS: 924-50-5 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00043940 InChI Key: FZIBCCGGICGWBP-UHFFFAOYSA-N Synonym: methyl 3-methyl-2-butenoate,methyl 3,3-dimethylacrylate,2-butenoic acid, 3-methyl-, methyl ester,3-methyl-but-2-enoic acid methyl ester,methyl senecioate,3,3-dimethylacrylic acid methyl ester,methyl-3,3-dimethylacrylate,crotonic acid, 3-methyl-, methyl ester,methyl-3,3-dimethyl acrylate,methylsenecioate PubChem CID: 13546 IUPAC Name: methyl 3-methylbut-2-enoate SMILES: CC(=CC(=O)OC)C