missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloro-6-(trifluoromethyl)nicotinate, TRC

Product Code. 30470206
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470206 10 mg 10mg
30469040 50 mg 50mg
30476689 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470206 Supplier Toronto Research Chemicals Supplier No. TRCM34052010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073129-57-3
Molecular Formula C8H5NO2F3Cl
Molecular Weight (g/mol) 239.58
IUPAC Name methyl 2-chloro-6-(trifluoromethyl)pyridine-3-carboxylate
SMILES COC(=O)c1ccc(nc1Cl)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H5ClF3NO2/c1-15-7(14)4-2-3-5(8(10,11)12)13-6(4)9/h2-3H,1H3
Formula Weight 238.9961
Chemical Name or Material Methyl 2-Chloro-6-(trifluoromethyl)nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.