CAS RN 1073129-57-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-Chloro-6-(trifluoromethyl)nicotinate, TRC
CAS: 1073129-57-3 Molecular Formula: C8H5NO2F3Cl Molecular Weight (g/mol): 239.58 IUPAC Name: methyl 2-chloro-6-(trifluoromethyl)pyridine-3-carboxylate SMILES: COC(=O)c1ccc(nc1Cl)C(F)(F)F