missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-bis(4-Hydroxyphenyl)acetate, TRC

Product Code. 30137169
Change view
Click to view available options
Quantity:
500 mg
2.5 g
5 g
Unit Size:
2.5g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137169 5 g 5g
30136050 2.5 g 2.5g
17920325 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137169 Supplier Toronto Research Chemicals Supplier No. TRCM2941655G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5129-00-0
Molecular Formula C15 H14 O4
Molecular Weight (g/mol) 258.27
Synonym Acetic acid, bis(p-hydroxyphenyl)-, methyl ester (6CI,7CI,8CI), Methyl 2,2-bis(4-hydroxyphenyl)acetate, Methyl bis(4-hydroxyphenyl)acetate, Methyl bis(p-hydroxyphenyl)acetate
IUPAC Name methyl 2,2-bis(4-hydroxyphenyl)acetate
SMILES COC(=O)C(c1ccc(O)cc1)c2ccc(O)cc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H14O4/c1-19-15(18)14(10-2-6-12(16)7-3-10)11-4-8-13(17)9-5-11/h2-9,14,16-17H,1H3
Formula Weight 258.0892
Percent Purity >95%
Chemical Name or Material Methyl 2,2-bis(4-Hydroxyphenyl)acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.