CAS RN 5129-00-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2,2-bis(4-Hydroxyphenyl)acetate, TRC
CAS: 5129-00-0 Molecular Formula: C15 H14 O4 Molecular Weight (g/mol): 258.27 Synonym: Acetic acid, bis(p-hydroxyphenyl)-, methyl ester (6CI,7CI,8CI),Methyl 2,2-bis(4-hydroxyphenyl)acetate,Methyl bis(4-hydroxyphenyl)acetate,Methyl bis(p-hydroxyphenyl)acetate IUPAC Name: methyl 2,2-bis(4-hydroxyphenyl)acetate SMILES: COC(=O)C(c1ccc(O)cc1)c2ccc(O)cc2