missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%

Product Code. 15476217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476217 5 g 5g
15466217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476217 Supplier Thermo Scientific Alfa Aesar Supplier No. H33407.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122684-33-7 | C11H19NO4 | 229.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122684-33-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD04038683
InChI Key LIWFYAVKYUQMRE-UHFFFAOYSA-N
Synonym methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748
PubChem CID 4572775
IUPAC Name 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC

Specifications

Boiling Point 105°C (0.2 mmHg)
Quantity 5 g
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material Methyl 1-Boc-pyrrolidine-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.