accessibility menu, dialog, popup

UserName

CAS RN 122684-33-7

CH3H3CCH3OONOOH3C

CAS RN 122684-33-7

IUPAC Name: 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate
Synonyms: 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: LIWFYAVKYUQMRE-UHFFFAOYNA-N
SMILES: COC(=O)C1CCN(C1)C(=O)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results