missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Sulfoxide Reductase A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57727
This item is not returnable.
View return policy
Description
Methionine Sulfoxide Reductase A Polyclonal specifically detects Methionine Sulfoxide Reductase A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Methionine Sulfoxide Reductase A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cytosolic methionine-S-sulfoxide reductase, EC 1.8.4.11, methionine sulfoxide reductase A, peptide met (O) reductase, Peptide Met(O) reductase, peptide methionine sulfoxide reductase, Peptide-methionine (S)-S-oxide reductase, PMSR, Protein-methionine-S-oxide reductase | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4482 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q9UJ68 | |
| MSRA | |
| Synthetic peptides corresponding to MSRA (methionine sulfoxide reductase A) The peptide sequence was selected from the middle region of MSRA)(50ug). Peptide sequence YQKVLSEHGFGPITTDIREGQTFYYAEDYHQQYLSKNPNGYCGLGGTGVS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Bovine: 92%. | |
| Human, Mouse, Rat, Pig, Canine, Equine, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction