Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-413
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
Brand:  Brady™ 104179
 View more versions of this product
Product Code. 11579423
  • ONLINE EXCLUSIVE PRICE
    £402.00
    £383.51 / 1 rolls
    Save £18.49
    4% Off
Estimated Shipment: 30-11-2026
Add to basket
Description
Description
  • Available in various sizes
  • Acrylic-based, industrial-strength adhesive for multiple applications
Specifications
Specifications
UL Recognised; cUL Recognised
Acrylic, Permanent
Light Gray
Roll
Single-side Printable
Label
Asset Tracking, General and Industrial Identificationentification, Rating Plates
1
THT-55-413-1
1000/Pk.
101.60 mm (W) x 50.80 mm (H)
-94°F to +230°F
0.0034 in.
53.98 mm
2 in.
101.60 mm
2 in.
THT-55-413-1
Thermal Transfer Printable Labels
None
Satin
Polyester (B-480)
Thermal Transfer
Non-sterile
BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
-70°C
1000 Label(s) / Roll
Rectangle
101.6 x 50.8 mm (W x H)
-70°C to +110°C
0.086 mm
50.8 x 101.60 mm
4 in.
101.60 mm
50.8 mm